Learn More
Simport™ Scientific Slimsette™ Tissue Processing/Embedding Cassettes in Quickload™ Stacks
Catalog No. 22038537 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-038-537 Lilac
22-038-530 Pink
22-038-531 Green
22-038-533 Blue
22-038-534 Peach
22-038-535 Tan
22-038-536 Gray
22-038-538 Orange
22-038-539 Aqua
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 22-038-537 Supplier Simport™ Scientific Supplier No. M50910T
Add to Cart
Add to Cart
This item is not returnable. View return policy
Secure specimens during processing with Slimsette™ Tissue Processing/Embedding Cassettes in Quickload™ Stacks.
  • In Quickload™ stacks
  • 45° Anterior writing area
  • Recessed cover
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Quantity 2000/Pk.
Color Lilac
Material Acetal