Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Safety-Walk™ Slip Resistant General Purpose Tape, Series 600

Catalog No. 17623910 Shop All 3M Products
Click to view available options
Color:
Black
Yellow
Width (Metric):
19 mm
25 mm
51 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Mineral coated, slip resistant material features a highly durable surface. Low profile design helps reduce tripping hazards.

  • Anti-slip tape for light to heavy shoe-traffic areas
  • High quality abrasive particles to ensure long-lasting friction in any application
  • Highly durable surface withstands the rigors of equipment traffic
  • High performance, pressure-sensitive adhesive for easy application
  • Custom shapes and sizes are also available
  • NFSI High Traction Certification
TRUSTED_SUSTAINABILITY

Specifications

Finish Mineral-coated
Adhesive Type Pressure Sensitive Adhesive
Dimensions (L x W) 18.3 m x 51 mm
Type Floor Tape
For Use With (Application) Light to Heavy Traffic Areas
Length (English) 60 ft.
Length (Metric) 18.3 m
Packaging Quantity 1 Ea.
Width (English) 2 in.
Width (Metric) 51 mm
Color Yellow
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.