Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLIRP Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11037258MFV450
Description
SLIRP Polyclonal antibody specifically detects SLIRP in Human, Mouse, Primate samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
SLIRP | |
Polyclonal | |
Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
chromosome 14 open reading frame 156, DC50, PD04872, SLIRP, SRA stem-loop-interacting RNA-binding protein, mitochondrial | |
A synthetic peptide made to a C-terminal region of human SLIRP protein. [Entrez-Prot# AAX58600] | |
0.1 mL | |
Breast Cancer, Cancer, Chromatin Research, DNA replication Transcription Translation and Splicing, Signal Transduction, Stem Cell Markers | |
81892 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction