Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLIRP Polyclonal Antibody
SLIRP Polyclonal Antibody
Supplier: Thermo Scientific PA5115923
Description
SLIRP Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)

Specifications
SLIRP | |
Polyclonal | |
Unconjugated | |
SLIRP | |
1810035L17Rik; C14orf156; DC23; DC50; PD04872; SLIRP; SRA stem-loop interacting RNA binding protein; SRA stem-loop-interacting RNA-binding protein, mitochondrial | |
Rabbit | |
Affinity Chromatography | |
RUO | |
380773, 688717, 81892 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q9D8T7, Q9GZT3 | |
SLIRP | |
A synthesized peptide derived from human SLIRP(Accession Q9GZT3), corresponding to amino acid residues Q79-F109. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction