Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLIT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121598
This item is not returnable.
View return policy
Description
SLIT1 Polyclonal Antibody for Western Blot, IHC (P)
SLIT1 is thought to act as molecular guidance cue in cellular migration, and function appears to be mediated by interaction with roundabout homolog receptors.
Specifications
SLIT1 | |
Polyclonal | |
Unconjugated | |
SLIT1 | |
KIAA0813; Megf4; mKIAA0813; Multiple EGF-like domains protein 4; multiple epidermal growth factor-like domains 4; multiple epidermal growth factor-like domains protein 4; SLIL1; slit guidance ligand 1; slit homolog 1; slit homolog 1 (Drosophila); slit homolog 1 protein; SLIT1; Slit-1; SLIT3 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
100627880, 65047, 6585 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
O75093, O88279 | |
SLIT1 | |
Recombinant protein Slit Homolog 1. The antigen corresponds to amino acid range 647-835 of the target protein. | |
20 μL | |
Primary | |
Human, Pig, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction