Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Slit2 Antibody (710305), DyLight 594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB5444M
Description
Slit2 Monoclonal antibody specifically detects Slit2 in Human, Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
Slit2 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry, CyTOF | |
710305 | |
Flow Cytometry, CyTOF-ready | |
FLJ14420, slit (Drosophila) homolog 2, slit homolog 2 (Drosophila), slit homolog 2 protein, Slit-2SLIL3 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Slit2, Gln26-Gln900, Accession # Q9R1B9 | |
0.1 mL | |
Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
9353 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction