Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLIT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115924
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115924 Supplier Invitrogen™ Supplier No. PA5115924
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLIT2.

SLIT2 are ligands for Robo receptors and play a vital role for axonal migration/navigation during neural development. A number of cleavage products through alternate splicing are reported in the literature for SLIT2 proteins. The C-terminal cleavage proteins are more diffusible than the larger N-terminal protein that is more tightly cell associated. SLIT2 protein is expressed in tissues such as fetal lung, kidney, and adult spinal cord.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLIT2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene SLIT2
Gene Accession No. O94813, Q9WVC1
Gene Alias downregulated during adipocyte differentiation-1; Drad-1; E030015M03Rik; E130320P19Rik; mKIAA4141; neurogenic extracellular slit protein; SLIL3; Slit; slit guidance ligand 2; slit homolog 2 (Drosophila); slit homolog 2 protein; Slit homolog 2 protein C-product; Slit homolog 2 protein N-product; SLIT2; Slit-2; Slit2 protein
Gene Symbols SLIT2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLIT2(Accession O94813), corresponding to amino acid residues N465-T515.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360272, 9353
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.