Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLIT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104142
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104142 Supplier Invitrogen™ Supplier No. PA5104142
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLIT3.

Slit guidance ligand 3 (SLIT3) is a gene located on chromosome 5q35.1, encoding a member of the SLIT family of proteins that play key roles in axon guidance and neuronal migration during development. SLIT3 functions as a secreted protein, influencing the directional growth and pathfinding of axons by interacting with its receptor, Robo, on the surface of neurons. This interaction is critical for establishing neural network architecture and maintaining proper neural connectivity. Beyond its role in the nervous system, SLIT3 is involved in processes such as angiogenesis and cell migration, impacting tissue development and repair. Dysregulation of SLIT3 expression or function has been associated with developmental abnormalities and various diseases, including cancer, where it may influence tumor cell invasion and metastasis. Research into SLIT3 continues to explore its therapeutic potential in modulating axon guidance and its implications in pathological conditions affecting both the nervous and vascular systems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLIT3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLIT3
Gene Accession No. O75094, O88280, Q9WVB4
Gene Alias b2b2362.1Clo; KIAA0814; Megf5; Multiple EGF-like domains protein 5; multiple epidermal growth factor-like domains 5; multiple epidermal growth factor-like domains protein 5; SLIL2; SLIL2FLJ10764; slit guidance ligand 3; slit homolog 3; slit homolog 3 (Drosophila); slit homolog 3 protein; SLIT1; slit2; SLIT3; Slit-3; UNQ691/PRO1336
Gene Symbols SLIT3
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLIT3(Accession O75094), corresponding to amino acid residues K1260-R1310.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20564, 6586, 83467
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.