Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLIT3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | SLIT3 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5121597
|
Invitrogen™
PA5121597 |
20 μL |
Each for $209.50
|
|
|||||
PIPA5121596
|
Invitrogen™
PA5121596 |
100 μL |
Each for $511.50
|
|
|||||
Description
SLIT3 may act as molecular guidance cue in cellular migration, and function may be mediated by interaction with roundabout homolog receptors.Specifications
SLIT3 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O75094 | |
6586 | |
Lys1405-Glu1512 (Accession O75094), with N-terminal His Tag. | |
Primary | |
-20°C | |
SLIT3 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
b2b2362.1Clo; KIAA0814; Megf5; Multiple EGF-like domains protein 5; multiple epidermal growth factor-like domains 5; multiple epidermal growth factor-like domains protein 5; SLIL2; SLIL2FLJ10764; slit guidance ligand 3; slit homolog 3; slit homolog 3 (Drosophila); slit homolog 3 protein; SLIT1; slit2; SLIT3; Slit-3; UNQ691/PRO1336 | |
SLIT3 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title