Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLIT3Rabbit anti-Bovine, Canine, Human, Mouse, Rat, Polyclonal, Rockland™
SDP

Catalog No. 501991840
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1840 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1840 Supplier Rockland Immunochemicals Supplier No. 600401884
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human SLIT-3 protein. A BLAST analysis was used to suggest reactivity with this protein from human, rat, dog, bovine and mouse based on 100% homology for the immunogen sequence. Expect cross reactivity with SLIT-3 from chimpanzee and chicken sources, as only a single amino acid residue change is found within the immunogen sequence (93% positive by BLAST). Cross reactivity with SLIT-3 homologues from other sources has not been determined.

SLIT3 may act as molecular guidance cue in cellular migration, and function may be mediated by interaction with roundabout homolog receptors.

Specifications

Antigen SLIT3
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene SLIT3
Gene Accession No. O75094, O88280, Q9WVB4
Gene Alias b2b2362.1Clo; BOS_19615; KIAA0814; Megf5; Multiple EGF-like domains protein 5; multiple epidermal growth factor-like domains 5; Multiple epidermal growth factor-like domains protein 5; SLIL2; SLIL2FLJ10764; slit guidance ligand 3; slit homolog 3; slit homolog 3 (Drosophila); Slit homolog 3 protein; SLIT1; slit2; Slit3; Slit-3; UNQ691/PRO1336
Gene Symbols SLIT3
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20564, 489132, 615883, 6586, 83467
Target Species Bovine, Canine, Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.