Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLITRK1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149541C
Description
SLITRK1 Polyclonal specifically detects SLITRK1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
SLITRK1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Q96PX8 | |
SLITRK1 | |
A genomic peptide made to an internal region of the human Slitrk1 protein (within residues 250-400). [Swiss-Prot Q96PX8] | |
0.1 mL | |
Neuroscience | |
114798 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FLJ54428, KIAA1910KIAA0918, leucine rich repeat containing 12, Leucine-rich repeat-containing protein 12, LRRC12, RP11-395N17.1, SLIT and NTRK-like family, member 1, SLIT and NTRK-like protein 1, slit and trk like gene 1, TTM | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human and mouse. Immunogen sequence has 84% homology to rat. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction