Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLITRK1 Recombinant Rabbit Monoclonal Antibody (1)
SLITRK1 Recombinant Rabbit Monoclonal Antibody (1)
Supplier: Thermo Scientific MA530887
Description

Specifications
SLITRK1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q96PX8 | |
SLITRK1 | |
Recombinant Human SLITRK1 protein (Met1-Ser616). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
1 | |
Unconjugated | |
SLITRK1 | |
3200001I04Rik; KIAA1910; Leucine-rich repeat-containing protein 12; LRRC12; RP11-395N17.1; SLIT and NTRK like family member 1; SLIT and NTRK-like family, member 1; SLIT and NTRK-like protein 1; slit and trk like gene 1; SLITRK1; Sltk1; TTM; UNQ233/PRO266 | |
Rabbit | |
Protein A | |
RUO | |
114798 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction