Learn More
SLITRK2 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5111909
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)KASRPPK MRNRPTPR Homology: Rat, human amino acid residues are identical.
This intronless gene is expressed in the hippocampus and maps close to a candidate region for several X-linked mental retardation (XLMR) syndromes. It is conserved in primates, cow, and horse, but not found in mouse and rat. The exact function of this gene is not known, but on the basis of its physical location and expression pattern, it is proposed to have an important function in the brain. [provided by RefSeq, Jan 2010].
Specifications
SLITRK2 (extracellular) | |
Polyclonal | |
Unconjugated | |
SLITRK2 | |
A930040J07; CXorf2; KIAA1854; RGD1561361; SLIT and NTRK-like protein 2; SLITL1; SLITRK2; UNQ9197/PRO34756 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
245450, 309349, 84631 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
Q810C0, Q9H156 | |
SLITRK2 | |
Peptide corresponding to amino acid residues 301-315 of mouse Slitrk2 (Extracellular, N-terminus) | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.