Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLITRK2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981818
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1818 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1818 Supplier Bioss Supplier No. BS11955R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This intronless gene is expressed in the hippocampus and maps close to a candidate region for several X-linked mental retardation (XLMR) syndromes. It is conserved in primates, cow, and horse, but not found in mouse and rat. The exact function of this gene is not known, but on the basis of its physical location and expression pattern, it is proposed to have an important function in the brain. [provided by RefSeq, Jan 2010].

Specifications

Antigen SLITRK2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SLITRK2
Gene Accession No. Q9H156
Gene Alias A930040J07; CXorf2; KIAA1854; RGD1561361; SLIT and NTRK like family member 2; SLIT and NTRK-like family, member 2; SLIT and NTRK-like protein 2; slit and trk like gene 2; SLITL1; slit-like 1; SLITRK2; UNQ9197/PRO34756
Gene Symbols SLITRK2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SLITRK2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84631
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.