Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLITRK4 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300076APC
Description
SLITRK4 Polyclonal antibody specifically detects SLITRK4 in Human samples. It is validated for ELISASpecifications
SLITRK4 | |
Polyclonal | |
ELISA | |
DKFZp547M2010, SLIT and NTRK-like family, member 4, SLIT and NTRK-like protein 4, slit and trk like gene 4 | |
Produced in rabbits immunized with purified, recombinant Human SLITRK4 (Uniprot#: Q8IW52; Met1-Pro616) | |
0.1 mL | |
Neuroscience | |
139065 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction