Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLITRK4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520481
Description
A suggested positive control is mouse brain tissue lysate. PA5-20481 can be used with blocking peptide PEP-0601.
SLIT and NTRK-like family 4 (Slitrk4) is a member a protein family consisting of six homologous transmembrane proteins (Slitrk1-6) that share two conserved leucine-rich repeat domains in the extracellular domain and have significant homology to Slit, a secreted axonal growth-controlling protein. These proteins are also homologous to trk neurotrophin receptors in their intracellular domains. Expression of Slitrk proteins is highly restricted to neural and brain tumor tissues, but varies within the protein family. Like every other Slitrk protein except Slitrk1, overexpression of Slitrk4 inhibited neurite outgrowth in cultured neurons, suggesting that these proteins are involved in the control of neurite outgrowth.
Specifications
| SLITRK4 | |
| Polyclonal | |
| Unconjugated | |
| SLITRK4 | |
| 6430513G13; D930039I09Rik; SLIT and NTRK like family member 4; SLIT and NTRK-like family, member 4; SLIT and NTRK-like protein 4; slit and trk like gene 4; Slitrk4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 139065, 245446, 302473 | |
| 4°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q810B8, Q8IW52 | |
| SLITRK4 | |
| A 13 amino acid peptide from near the carboxy terminus of human Slitrk4. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction