Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLITRK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581873
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581873 Supplier Invitrogen™ Supplier No. PA581873
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PDCGSMQLQL RKHDHKTNKK DGLSTEAFIP QTIEQMSKSH ACGLKESETG FMFSDPPGQK VVMRNVADKE KDLLHVDTRK RLSTIDELDE LFPSRDSNVF IQNFLESKKE YNSIGVSGFE IRYPEKQPDK KSKKSLIGGN HS Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 24%.

SLIT and NTRK-like family 4 (Slitrk4) is a member a protein family consisting of six homologous transmembrane proteins (Slitrk1-6) that share two conserved leucine-rich repeat domains in the extracellular domain and have significant homology to Slit, a secreted axonal growth-controlling protein. These proteins are also homologous to trk neurotrophin receptors in their intracellular domains. Expression of Slitrk proteins is highly restricted to neural and brain tumor tissues, but varies within the protein family. Like every other Slitrk protein except Slitrk1, overexpression of Slitrk4 inhibited neurite outgrowth in cultured neurons, suggesting that these proteins are involved in the control of neurite outgrowth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLITRK4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLITRK4
Gene Accession No. Q8IW52
Gene Alias 6430513G13; D930039I09Rik; SLIT and NTRK like family member 4; SLIT and NTRK-like family, member 4; SLIT and NTRK-like protein 4; slit and trk like gene 4; Slitrk4
Gene Symbols SLITRK4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLITRK4. Recombinant protein control fragment (Product #RP-92105).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139065
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.