Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLITRK6 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300225PCP
Description
SLITRK6 Polyclonal antibody specifically detects SLITRK6 in Human samples. It is validated for ELISASpecifications
SLITRK6 | |
Polyclonal | |
ELISA | |
FLJ22774, MGC119595, MGC119596, MGC119597,4832410J21Rik, SLIT and NTRK-like family, member 6, SLIT and NTRK-like protein 6, slit and trk like gene 6 | |
Produced in rabbits immunized with purified, recombinant Human SLITRK6 (Uniprot#: Q9H5Y7; Met1-Ser608) | |
0.1 mL | |
Neuroscience | |
84189 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction