Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLITRK6 Polyclonal Antibody

SLITRK6 Polyclonal Antibody
Supplier: Thermo Scientific PA533054
Description
SLITRK6 Polyclonal Antibody for IHC (P)
Specifications
| SLITRK6 | |
| Polyclonal | |
| Unconjugated | |
| SLITRK6 | |
| 4832410J21Rik; DFNMYP; neuronal transmembrane protein Slitrk6; RP11-272M24.2; SLIT and NTRK like family member 6; SLIT and NTRK-like family, member 6; SLIT and NTRK-like protein 6; slit and trk like gene 6; SLITRK6; Sltk6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100153917, 100856466, 781119, 84189 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9H5Y7 | |
| SLITRK6 | |
| Synthetic 14 amino acid peptide from internal region of human SLITRK6. | |
| 50 μg | |
| Primary | |
| Human, Canine, Monkey, Bovine, Pig, Bat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction