Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLK Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298482AF647
Description
SLK Polyclonal specifically detects SLK in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| SLK | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| bA16H23.1, CTCL tumor antigen se20-9, EC 2.7.11, EC 2.7.11.1, hSLK, KIAA0204LOSK, Long Ste20-like Kinase, se20-9, Serine/threonine-protein kinase 2, SNF1 (sucrose nonfermenting, yeast, homolog)-like kinase, SNF1 sucrosenonfermenting like kinase, SNF1 (sucrose nonfermenting, yeast, homolog)-like kinase, SNF1 sucrosenonfermenting like kinase (yeast), SNF1 sucrose nonfermenting like kinase, STE20-like kinaseSTE20-like kinase (yeast), STE20-like serine/threonine-protein kinase, STE20-related kinase, Ste20-related serine/threonine kinase, STE20-related serine/threonine-protein kinase, STK2MGC133067 | |
| Produced in rabbits immunized with E. coli-derived Human SLK fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 9748 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction