Learn More
Promega SLK Kinase Enzyme System, 10microg
Catalog No. PRV4242 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4242 10 μg
1 options
Catalog No. PRV4242 Supplier Promega Supplier No. V4242
Recombinant full-length human SLK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. SLK or LOSK is a Ste20-like protein kinase that associates with microtubule and centrosome.
  • Easily Screen and Profile SLK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C