Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580649
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580649 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580649 Supplier Invitrogen™ Supplier No. PA580649
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human SLK.

SLK acts as a novel effector of cytoskeletal remodeling and apoptosis, and seems to be ubiquitously expressed. SLK mediates apoptosis and actin stress fiber dissolution.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLK
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SLK
Gene Accession No. Q9H2G2
Gene Alias 9A2; AV021402; AW411554; bA16H23.1; CTCL tumor antigen se20-9; Etk4; hSLK; Kiaa0204; Long Ste20-like Kinase; LOSK; mKIAA0204; mSLK; se20-9; serine/threonine kinase 2; serine/threonine-protein kinase 2; Sk2; Slk; SMAK; SNF1 (sucrose nonfermenting, yeast, homolog)-like kinase, SNF1 sucrose nonfermenting like kinase; SNF1 sucrose nonfermenting like kinase; STE20 like kinase; STE20-like kinase; STE20-like serine/threonine-protein kinase; STE20-related kinase; STE20-related kinase SMAK; Ste20-related serine/threonine kinase; STE20-related serine/threonine-protein kinase; Stk2
Gene Symbols SLK
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human SLK.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9748
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.