Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLK Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985464
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5464 Supplier Bioss Supplier No. BS3519R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLK mediates apoptosis and actin stress fiber dissolution.

Specifications

Antigen SLK
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SLK
Gene Accession No. Q9H2G2
Gene Alias 9A2; AV021402; AW411554; bA16H23.1; CTCL tumor antigen se20-9; Etk4; hSLK; KIAA0204; Long Ste20-like Kinase; LOSK; mKIAA0204; mSLK; se20-9; serine/threonine kinase 2; Serine/threonine-protein kinase 2; Sk2; SLK; SMAK; SNF1 (sucrose nonfermenting, yeast, homolog)-like kinase, SNF1 sucrose nonfermenting like kinase; SNF1 sucrose nonfermenting like kinase; STE20 like kinase; STE20-like kinase; STE20-like serine/threonine-protein kinase; STE20-related kinase; STE20-related kinase SMAK; Ste20-related serine/threonine kinase; STE20-related serine/threonine-protein kinase; STK2
Gene Symbols SLK
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SLK.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9748
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.