Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLMAP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$405.00 - $670.00
Specifications
Antigen | SLMAP |
---|---|
Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SLMAP Polyclonal specifically detects SLMAP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SLMAP | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
7871 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:SEYEKEITSLQNSFQLRCQQCEDQQREEATRLQGELEKLRKEWNALETECHSLKRENVLLSSELQRQEKELHNSQKQSLELTSDLSILQMSRKELENQVGSLKEQHLRDSADLKTLLSKAENQAKDVQKEYEKTQTVLSELKLKFEMTEQ | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 | |
Polyclonal | |
Rabbit | |
Mouse, Human, Rat | |
FLJ42206, KIAA1601Sarcolemmal-associated protein, MGC138760, sarcolemma associated protein, sarcolemmal membrane-associated protein, SLAPMGC138761 | |
SLMAP | |
IgG | |
Affinity Purified | |
Specificity of human SLMAP antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title