Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLMAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551708
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551708 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551708 Supplier Invitrogen™ Supplier No. PA551708
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SEYEKEITSL QNSFQLRCQQ CEDQQREEAT RLQGELEKLR KEWNALETEC HSLKRENVLL SSELQRQEKE LHNSQKQSLE LTSDLSILQM SRKELENQVG SLKEQHLRDS ADLKTLLSKA ENQAKDVQKE YEKTQTVLSE LKLKFEMTEQ Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.

Sarcolemmal membrane associated protein (SLMAP) is a tail-anchored membrane protein involved in the docking and membrane fusion process via protein-protein interactions. Deregulation of SLMAP expression plays an important role in type 2 diabetes, and changes in SLMAP expression are linked to hyperglycemia and diabetes. SLMAPs are components of cardiac membranes and have been identified as a novel regulator of cardiac function. There are several isoforms of SLMAPs with varying molecular size (35, 45, 63, and 83-91 kDa) as a result of alternative splicing. The short 35- kDa isoform is predominantly expressed in cardiac and slow-twitch skeletal muscle, while the 83- to 91- kDa isoform is ubiquitous.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLMAP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Slmap
Gene Accession No. P0C219, Q14BN4
Gene Alias D330001L02Rik; KIAA1601; mKIAA1601; sarcolemma associated protein; sarcolemmal membrane-associated protein; Sarcolemmal-associated protein; Slap; Slmap; UNQ1847/PRO3577
Gene Symbols Slmap
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLMAP. Recombinant protein control fragment (Product #RP-102135).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 290533, 7871
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.