Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLMO2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548369
Description
Predicted to react with bovine based on sequence homology.
PRELID3B gene ontology annotations related to this gene include phospholipid transport.
Specifications
SLMO2 | |
Polyclonal | |
Unconjugated | |
PRELID3B | |
2310042G06Rik; AA407013; C20orf45; CGI-107; CGI-107 protein; dJ543J19.5; hypothetical protein LOC616523; likely orthologue of H. sapiens chromosome 20 open reading frame 45 (C20orf45); PRELI domain containing 3B; PRELI domain containing protein 3B; Prelid3b; Protein slowmo homolog 2; SLMO2; slowmo homolog 2; slowmo homolog 2 (Drosophila); slowmo-like protein 2-like protein | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
51012 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9Y3B1 | |
PRELID3B | |
KLH conjugated synthetic peptide between 92-121 amino acids from the Central region of human SLMO2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction