Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLMO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570245
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570245 Supplier Invitrogen™ Supplier No. PA570245
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

PRELID3B gene ontology annotations related to this gene include phospholipid transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLMO2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PRELID3B
Gene Accession No. Q9Y3B1
Gene Alias 2310042G06Rik; AA407013; C20orf45; CGI-107; CGI-107 protein; dJ543J19.5; hypothetical protein LOC616523; likely orthologue of H. sapiens chromosome 20 open reading frame 45 (C20orf45); PRELI domain containing 3B; PRELI domain containing protein 3B; Prelid3b; Protein slowmo homolog 2; SLMO2; slowmo homolog 2; slowmo homolog 2 (Drosophila); slowmo-like protein 2-like protein
Gene Symbols PRELID3B
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human SLMO2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51012
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.