Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLMO2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA561575
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561575 Supplier Thermo Scientific Supplier No. PA561575
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASTISSNASK GREAMEWVIH KLNAEIEELT ASARGTIRTP MAAAAFAEK Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.

PRELID3B gene ontology annotations related to this gene include phospholipid transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLMO2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRELID3B
Gene Accession No. Q9Y3B1
Gene Alias 2310042G06Rik; AA407013; C20orf45; CGI-107; CGI-107 protein; dJ543J19.5; hypothetical protein LOC616523; likely orthologue of H. sapiens chromosome 20 open reading frame 45 (C20orf45); PRELI domain containing 3B; PRELI domain containing protein 3B; Prelid3b; Protein slowmo homolog 2; SLMO2; slowmo homolog 2; slowmo homolog 2 (Drosophila); slowmo-like protein 2-like protein
Gene Symbols PRELID3B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLMO2
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51012
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.