Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLN Monoclonal Antibody (14G1A12)
GREENER_CHOICE

Catalog No. FEMA550293
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550293 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550293 Supplier Invitrogen™ Supplier No. MA550293
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLN Monoclonal Antibody for IHC (P), Flow, ELISA

SLN, also named as Sarcolipin, belongs to the sarcolipin family. It is associated with calcium ATPase SERCA1. It inhibits SERCA pumps. SLN, a key regulator of cardiac sarco (endo)plasmic reticulum (SR) Ca(2+) ATPase, is predominantly expressed in atria and mediates beta-adrenergic responses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLN
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 14G1A12
Concentration 1.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Sln
Gene Accession No. O00631
Gene Alias 2310045A07Rik; Sarcolipin; SLN
Gene Symbols Sln
Host Species Mouse
Immunogen Recombinant Human SLN protein (1-31AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6588
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.