Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLN13 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA554599
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554599 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554599 Supplier Thermo Scientific Supplier No. PA554599

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Immunogen sequence: VLGCAKEQVD PDSLKNVIAR AISKLPIVHF CSSKPRVEYS TKIVEVFCGK ELYGYLCVIK VKAFCCVVFS EAPK Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 65%.

SLN13 Polyclonal antibody specifically detects SLN13 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLN13
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLFN13
Gene Accession No. Q68D06
Gene Alias hSLFN13; Schlafen family member 13; Schlafen-13; SLFN10; SLFN13
Gene Symbols SLFN13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLN13
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146857
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.