Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Slo2.2 Monoclonal Antibody (N3/26), FITC
GREENER_CHOICE

Catalog No. PIMA545435
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545435 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545435 Supplier Invitrogen™ Supplier No. MA545435
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45435 was sufficient for detection of Slo2.2 in 10 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 140kDa. Weak human detection.approximately Does not cross-react with KCNT2/Slo2.1/Slick.

Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a sodium-activated potassium channel subunit which is thought to function in ion conductance and developmental signaling pathways. Mutations in this gene cause the early-onset epileptic disorders, malignant migrating partial seizures of infancy and autosomal dominant nocturnal frontal lobe epilepsy. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Slo2.2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Microarray
Classification Monoclonal
Clone N3/26
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene Kcnt1
Gene Accession No. Q5JUK3, Q6ZPR4, Q9Z258
Gene Alias bA100C15.2; C030030G16Rik; EIEE14; ENFL5; KCa4.1; KCa4.1KIAA1422FLJ41282; Kcnt1; KIAA1422; Potassium channel subfamily T member 1; potassium channel subunit (Slack); potassium channel, sodium activated subfamily T, member 1; potassium channel, sodium-activated subfamily T, member 1; potassium channel, subfamily T, member 1; potassium sodium-activated channel subfamily T member 1; rSlo2; Sequence like a calcium-activated K+ channel; sequence like a calcium-activated potassium channel subunit; Slack; slo2; Slo2.2
Gene Symbols Kcnt1
Host Species Mouse
Immunogen Fusion protein amino acids 1168-1237 of rat Slo2.2 (Slack).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227632, 57582, 60444
Target Species Human, Mouse, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.