Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Slow Skeletal Muscle Myosin Heavy Chain Antibody (9812), Novus Biologicals™
SDP

Catalog No. p-200057796 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395473 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB395473 Supplier Novus Biologicals Supplier No. NBP2502990.025ML
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Slow Skeletal Muscle Myosin Heavy Chain Monoclonal antibody specifically detects Slow Skeletal Muscle Myosin Heavy Chain in Human, Mouse, Rat, Chicken, Rabbit samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Slow Skeletal Muscle Myosin Heavy Chain
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 9812
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Formulation Ascites
Host Species Mouse
Immunogen Purified human skeletal muscle myosin
Purification Method Ascites
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cellular Signaling, Developmental Biology, Stem Cells
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Chicken, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.