Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Slow Skeletal Muscle Myosin Heavy Chain Antibody (M14) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | Slow Skeletal Muscle Myosin Heavy Chain |
---|---|
Clone | M14 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence |
Description
Slow Skeletal Muscle Myosin Heavy Chain Monoclonal antibody specifically detects Slow Skeletal Muscle Myosin Heavy Chain in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
Slow Skeletal Muscle Myosin Heavy Chain | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cardiovascular Biology, Cellular Signaling, Developmental Biology, Stem Cells | |
Ascites | |
IgG1 | |
Ascites |
M14 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
Monoclonal | |
Ascites | |
RUO | |
Human, Mouse, Rat, Chicken | |
Purified human skeletal muscle myosin | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title