Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLP-2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP17990320 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17990320 20 μL
NBP179903 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17990320 Supplier Novus Biologicals Supplier No. NBP17990320UL

Rabbit Polyclonal Antibody

SLP-2 Polyclonal specifically detects SLP-2 in Human samples. It is validated for Western Blot.

Specifications

Antigen SLP-2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_038470
Gene Alias EPB72-like protein 2, HSPC108, SLP2, SLP-2stomatin-like protein 2, stomatin (EPB72)-like 2, stomatin-like 2
Gene Symbols STOML2
Host Species Rabbit
Immunogen The specific Immunogen is proprietary information. Peptide sequence VTSMVAQAMGVYGALTKAPVPGTPDSLSSGSSRDVQGTDASLDEELDRVK.
Molecular Weight of Antigen 39 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 30968
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.