Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLP-2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP280112
Description
SLP-2 Polyclonal specifically detects SLP-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
SLP-2 | |
Polyclonal | |
Unconjugated | |
PBS | |
STOML2 | |
Synthetic peptide corresponding to aa 199-213 of mouse SLP-2. | |
100 μg | |
Core ESC Like Genes, Stem Cell Markers | |
30968 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence, Immunoprecipitation | |
1 mg/mL | |
Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
EPB72-like protein 2, HSPC108, SLP2, SLP-2stomatin-like protein 2, stomatin (EPB72)-like 2, stomatin-like 2 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Recognizes human, mouse and rat SLP-2. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction