Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLP-2 Antibody (OTI1F5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274227H
Description
SLP-2 Monoclonal specifically detects SLP-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SLP-2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
STOML2 | |
Full length recombinant protein of human SLP-2 (NP_038470) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1F5 | |
Western Blot, Immunohistochemistry | |
EPB72-like protein 2, HSPC108, SLP2, SLP-2stomatin-like protein 2, stomatin (EPB72)-like 2, stomatin-like 2 | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
30968.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction