Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLP-2 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298494PE
Description
SLP-2 Polyclonal antibody specifically detects SLP-2 in Human samples. It is validated for Western BlotSpecifications
SLP-2 | |
Polyclonal | |
Western Blot | |
EPB72-like protein 2, HSPC108, SLP2, SLP-2stomatin-like protein 2, stomatin (EPB72)-like 2, stomatin-like 2 | |
Produced in rabbits immunized with E. coli-derived Human SLP-2 fragment. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
30968 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction