Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLP-76/LCP2 Antibody (460004) [DyLight 405], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5637E
Description
SLP-76/LCP2 Monoclonal antibody specifically detects SLP-76/LCP2 in Human samples. It is validated for Western BlotSpecifications
SLP-76/LCP2 | |
Monoclonal | |
DyLight 405 | |
lymphocyte cytosolic protein 2, lymphocyte cytosolic protein 2 (SH2 domain containing leukocyte protein of76kDa), lymphocyte cytosolic protein 2 (SH2 domain-containing leukocyte protein of76kD), SH2 domain-containing leukocyte protein of 76 kDa, SH2 domain-containing leukocyte protein of 76kD, SLP-76, SLP-76 tyrosine phosphoprotein, SLP7676 kDa tyrosine phosphoprotein | |
E. coli-derived recombinant human SLP-76, Met1-Pro533, Accession # Q13094 | |
0.1 mL | |
Phospho Specific, Signal Transduction | |
3937 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot | |
460004 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction