Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ SLP76, CT, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABF1215 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABF1215 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF1215 Supplier MilliporeSigma™ Supplier No. ABF1215
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lymphocyte cytosolic protein 2 (UniProt Q13094; also known as SH2 domain-containing leukocyte protein of 76 kDa, SLP-76 tyrosine phosphoprotein, SLP76) is encoded by the LCP2 gene (Gene ID 3937) in human. SLP76 is a signaling adaptor molecule expressed in a variety of hematopoietic cells, including lymphocytes, myeloid cells, and platelets. SLP-76 contains three pYESP motifs (Y113 and Y128) that are phosphorylated by ZAP-70 to produce docking sites for SH2-containing proteins, such as exchange factor Vav1, adaptor Nck, resting lymphocyte kinase Rlk, and inducible tyrosine kinase Itk. In addition, SLP-76 binds to the SH3 domain of phospholipase Cγ1 (PLCγ1) and SLP76-deficient T cells show an impaired PLCγ1 activation and calcium mobilization. SLP-76 is essential for mediating TCR signaling, Lcp2-/- mice exhibit impaired thymic development and their T cells fail to develop beyond stage III at the double-negative stage. SLP76 is also a critical determinant of NK-cell development and the the underlying “missing-self” recognition mechanism that prevents NK cells from destroying cells that express self major histocompatibility complex (MHC) class I molecules. SLP76 contains an N-terminal sterile-α motif or SAM (human a.a. 15-81 and moust a.a. 12-78) and a C-terminal SH2 domain (human/mouse a.a. 422-530).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLP76, CT
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene LCP2
Gene Accession No. Q13094
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a central region sequence within the N-terminal half of human SLP76.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_005556
Test Specificity This rabbit polyclonal antibody targets a central region sequence within the N-terminal half of human SLP76.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.