Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLP76 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596165
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596165 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596165 Supplier Invitrogen™ Supplier No. PA596165
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-937, BT-474, Mouse spleen, Mouse liver; Cellular Location: Cytoplasm Immunogen sequence: SILPAKPFPN SNSMYIDRPP SGKTPQQPPV PPQRPMAALP PPPAGRNHSP LPPPQTNHEE PSRSRNHKTA KLPAPSIDRS TKPPLDRSLA PFDREPFTLG KKPPFSDKPS IPAGRSLGEH LPKIQKPPLP PTTERHERSS PLPGKKPPVP KHGWGPDRRE NDEDDVHQRP LPQPALLPMS SNTFPSRSTK PSPMNPLPSS HMPGAFSESN SSFPQSASLP PYFSQGPSNR PPIRAEGRNF PLPLPNKPRP PSPAEEENSL NEEWYVSYIT RPEAEAALRK INQDGTFLVR DSSKKTTTNP YVLMVLYKDK VYNIQIRYQK ESQVYLLGTG LRGKEDFLSV SDIIDYFRKM PLLLIDGKNR GSRYQCTLTH AAGYP.

SLP-76 was originally identified as a substrate of the ZAP-70 protein tyrosine kinase following T cell receptor (TCR) ligation in the leukemic T cell line Jurkat. The SLP-76 locus has been localized to human chromosome 5q33 and the gene structure has been partially characterized in mice. The human and murine cDNAs both encode 533 amino acid proteins that are 72% identical and comprised of three modular domains. The NH2-terminus contains an acidic region that includes a PEST domain and several tyrosine residues which are phosphorylated following TCR ligation. SLP-76 also contains a central proline-rich domain and a COOH-terminal SH2 domain. A number of additional proteins have been identified that associate with SLP-76 both constitutively and inducibly following receptor ligation, supporting the notion that SLP-76 functions as an adaptor or scaffold protein. Studies using SLP-76 deficient T cell lines or mice have provided strong evidence that SLP-76 plays a positive role in promoting T cell development and activation as well as mast cell and platelet function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLP76
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LCP2
Gene Accession No. Q13094, Q60787
Gene Alias 76 kDa tyrosine phosphoprotein; AI323664; BB161688; Lcp2; Lymphocyte cytosolic protein 2; lymphocyte cytosolic protein 2 (SH2 domain containing leukocyte protein of 76kDa); lymphocyte cytosolic protein 2 (SH2 domain-containing leukocyte protein of 76kD); SH2 domain-containing leukocyte protein of 76 kDa; SH2 domain-containing leukocyte protein of 76kD; SLP76; SLP-76; SLP-76 tyrosine phosphoprotein; twm
Gene Symbols LCP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 159-533 of human LCP2 (NP_0055561).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16822, 3937
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.