Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ SLP76, Rabbit, Alexa Fluor 488, Polyclonal,

Catalog No. ABF1215YMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 488
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABF1215YMI 100 μL Alexa Fluor 488
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF1215YMI Supplier MilliporeSigma™ Supplier No. ABF1215AF488

Rabbit Polyclonal Antibody

Lymphocyte cytosolic protein 2 (UniProt: Q13094; also known as Lymphocyte cytosolic protein 2, SH2 domain-containing leukocyte protein of 76 kDa, SLP-76 tyrosine phosphoprotein, SLP76) is encoded by the gene LCP2 (Gene ID 3937) in human. SLP-76 is a cytoplasmic adaptor phosphoprotein that contains both SH2 and SAM domains and is involved in B and T cell receptor signaling. Its amino terminus contains an acidic region that includes a PEST domain and several tyrosine residues that are phosphorylated following TCR ligation. SLP-76 also contains a central proline-rich domain and a SH2 domain at its carboxy terminal. SLP-76 can be phosphorylated on multiple tyrosine residues by the upstream kinases ZAP-70 and Lck. SLP-76 phosphorylation plays an important role in T cell-mediated IL-2 production by allowing phosphorylated Vav to bind and this complex stimulates NFAT and IL-2 gene activation after TCR engagement. SLP-76 is highly expressed in spleen, thymus, and peripheral blood leukocytes and T cells. It is expressed in B cell lines at lower levels and is not detected in fibroblast and neuroblastoma cell lines. SLP-76 is reported to play a positive role in promoting T cell development and activation as well as mast cell and platelet function. Overexpression of SLP-76 is shown to enhance IL-2 transcription after TCR signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLP76
Applications Flow Cytometry
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Alexa Fluor 488
Formulation Purified rabbit polyclonal antibody Alexa Fluor™ 488 conjugate in PBS with 1.5% BSA and 0.05% Sodium Azide.
Gene LCP2
Gene Accession No. Q13094
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to an internal sequence within the N-terminal half of human SLP76.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_005556
Test Specificity This rabbit polyclonal antibody Alexa Fluor™ 488 conjugate detected SLP76 (lymphocyte cytosolic protein 2) in Jurkat cells by flow cytometry.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.