Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLPI Antibody (001), DyLight 755 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289438IR
Description
SLPI Monoclonal antibody specifically detects SLPI in Human samples. It is validated for ELISA, ImmunofluorescenceSpecifications
SLPI | |
Monoclonal | |
DyLight 755 | |
ALK1, ALPProtease inhibitor WAP4, antileukoproteinase, BLPISeminal proteinase inhibitor, HUSI, HUSI-I, Mucus proteinase inhibitor, secretory leukocyte peptidase inhibitor, Secretory leukocyte protease inhibitor, secretory leukocyte protease inhibitor (antileukoproteinase), WAP four-disulfide core domain protein 4, WAP4HUSI-1, WFDC4MPI | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human SLPI (P03973; Met1-Ala132). | |
0.1 mL | |
Cancer | |
6590 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunofluorescence | |
001 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction