Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLPI Monoclonal Antibody (31)
GREENER_CHOICE

Catalog No. PIMA533381
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533381 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533381 Supplier Invitrogen™ Supplier No. MA533381
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Product is supplied at 1 mL (100 μL/mL). Application Notes: IHC-P - Specimen was fixed in 4% formalin for 4h. Paraffin sections (4 μm) were deparifinized, rehydrated and blocked with normal serum. Optional antigen retrieval with citrate buffer. Section was incubated with 5 μg/mL mAb. Immunoassay - This antibody can be used as coat and detection antibody. IHC-F - Permeabilization was done in cold acetone with 0.5% hydrogen peroxidase for 10 min, after drying and washing, antibodies were incubated for 30 minutes.

SLPI is a secreted inhibitor which protects epithelial tissues from serine proteases. It is found in various secretions including seminal plasma, cervical mucus, and bronchial secretions, and has affinity for trypsin, leukocyte elastase, and cathepsin G. Its inhibitory effect contributes to the immune response by protecting epithelial surfaces from attack by endogenous proteolytic enzymes; the protein is also thought to have broad-spectrum anti-biotic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLPI
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 31
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene SLPI
Gene Accession No. P03973
Gene Alias ALK1; ALP; antileukoproteinase; BLPI; HUSI; HUSI-1; HUSI-I; MPI; Mucus proteinase inhibitor; protease inhibitor WAP4; RP23-462O16.7; secretory leukocyte peptidase inhibitor; secretory leukocyte protease inhibitor; secretory leukocyte protease inhibitor (antileukoproteinase); seminal proteinase inhibitor; SLPI; WAP four-disulfide core domain protein 4; WAP4; WFDC4
Gene Symbols SLPI
Host Species Mouse
Immunogen SLPI purified from sputum.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6590
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.