Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLPI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582990
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582990 Supplier Invitrogen™ Supplier No. PA582990
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGKSFKAGVC PPKKSAQCLR YKKPECQSDW QCPGKKRCCP DTCGIKCLDP VDTPNPTRRK PGKCPVTYGQ CLMLNPPNFC EMDGQCKRDL KCCMGMCGKS CV Highest antigen sequence indentity to the following orthologs: Rat - 58%, Mouse - 55%.

SLPI is a secreted inhibitor which protects epithelial tissues from serine proteases. It is found in various secretions including seminal plasma, cervical mucus, and bronchial secretions, and has affinity for trypsin, leukocyte elastase, and cathepsin G. Its inhibitory effect contributes to the immune response by protecting epithelial surfaces from attack by endogenous proteolytic enzymes; the protein is also thought to have broad-spectrum anti-biotic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLPI
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLPI
Gene Accession No. P03973
Gene Alias ALK1; ALP; antileukoproteinase; BLPI; HUSI; HUSI-1; HUSI-I; MPI; Mucus proteinase inhibitor; protease inhibitor WAP4; RP23-462O16.7; secretory leukocyte peptidase inhibitor; secretory leukocyte protease inhibitor; secretory leukocyte protease inhibitor (antileukoproteinase); seminal proteinase inhibitor; SLPI; WAP four-disulfide core domain protein 4; WAP4; WFDC4
Gene Symbols SLPI
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLPI. Recombinant protein control fragment (Product #a href=.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6590
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.