Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLTM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559154
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559154 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559154 Supplier Invitrogen™ Supplier No. PA559154
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STDTPNKKPT KGKGKKHEAD ELSGDASVED DAFIKDCELE NQEAHEQDGN DELKDSEEFG ENEEENVHSK ELLSAEENKR AHELIEAEGI EDIEKEDIES QEIEAQEGED D Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.

SLTM gene ontology annotations related to this gene include RNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLTM
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLTM
Gene Accession No. Q9NWH9
Gene Alias 5730455C01Rik; 5730555F13Rik; 9130215G10Rik; FLJ13213; Met; modulator of estrogen induced transcription; modulator of estrogen-induced transcription; RGD1307526; SAFB like transcription modulator; SAFB-like; SAFB-like transcription modulator; SAFB-like, transcription modulator; SAF-like transcription modulator; SLTM
Gene Symbols SLTM
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLTM. Recombinant protein control fragment (Product #RP-102889).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79811
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.