Learn More
SLTM Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302835AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
SLTM gene ontology annotations related to this gene include RNA binding.Specifications
SLTM | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9NWH9 | |
SLTM | |
Between 800 and 850 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
SLTM | |
FLJ13213, Met, modulator of estrogen-induced transcription, SAFB-like, transcription modulator | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79811 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.