Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLU7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528513
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528513 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528513 Supplier Invitrogen™ Supplier No. PA528513
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T transfected SLU7 recombinant protein. Predicted reactivity: Mouse (90%), Rat (89%), Rhesus Monkey (98%), Chimpanzee (99%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Pre-mRNA splicing occurs in two sequential transesterification steps. The protein encoded by this gene is a splicing factor that has been found to be essential during the second catalytic step in the pre-mRNA splicing process. It associates with the spliceosome and contains a zinc knuckle motif that is found in other splicing factors and is involved in protein-nucleic acid and protein-protein interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLU7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene SLU7
Gene Accession No. O95391
Gene Alias 9G8; AU018913; D11Ertd730e; D3Bwg0878e; hSlu7; MGC9280; Pre-mRNA-splicing factor SLU7; SLU7; SLU7 homolog, splicing factor; SLU7 splicing factor homolog; SLU7 splicing factor homolog (S. cerevisiae); splicing factor; step II splicing factor SLU7; zinc knuckle motif containing
Gene Symbols SLU7
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human SLU7. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10569
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.