Learn More
Invitrogen™ SLU7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531127
Description
Recommended positive controls: A431, HeLa, HepG2, 293T transfected SLU7 recombinant protein. Predicted reactivity: Human (99%), Mouse (98%), Rat (97%), Zebrafish (80%), Chicken (86%), Rhesus Monkey (99%), Chimpanzee (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Pre-mRNA splicing occurs in two sequential transesterification steps. The protein encoded by this gene is a splicing factor that has been found to be essential during the second catalytic step in the pre-mRNA splicing process. It associates with the spliceosome and contains a zinc knuckle motif that is found in other splicing factors and is involved in protein-nucleic acid and protein-protein interactions.
Specifications
SLU7 | |
Polyclonal | |
Unconjugated | |
SLU7 | |
9G8; AU018913; D11Ertd730e; D3Bwg0878e; hSlu7; MGC9280; Pre-mRNA-splicing factor SLU7; SLU7; SLU7 homolog, splicing factor; SLU7 splicing factor homolog; SLU7 splicing factor homolog (S. cerevisiae); splicing factor; step II splicing factor SLU7; zinc knuckle motif containing | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10569, 193116 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
O95391, Q8BHJ9 | |
SLU7 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 282 of Human SLU7. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.