Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLU7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534916
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (100%), Rat (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Pre-mRNA splicing occurs in two sequential transesterification steps. The protein encoded by this gene is a splicing factor that has been found to be essential during the second catalytic step in the pre-mRNA splicing process. It associates with the spliceosome and contains a zinc knuckle motif that is found in other splicing factors and is involved in protein-nucleic acid and protein-protein interactions.
Specifications
SLU7 | |
Polyclonal | |
Unconjugated | |
SLU7 | |
9G8; AU018913; D11Ertd730e; D3Bwg0878e; hSlu7; MGC9280; Pre-mRNA-splicing factor SLU7; SLU7; SLU7 homolog, splicing factor; SLU7 splicing factor homolog; SLU7 splicing factor homolog (S. cerevisiae); splicing factor; step II splicing factor SLU7; zinc knuckle motif containing | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10569 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O95391 | |
SLU7 | |
Synthetic peptide corresponding to a region within amino acids 161 and 586 of Slu7. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction