Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLU7 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8327R

Description
Pre-mRNA splicing occurs in two sequential transesterification steps. The protein encoded by this gene is a splicing factor that has been found to be essential during the second catalytic step in the pre-mRNA splicing process. It associates with the spliceosome and contains a zinc knuckle motif that is found in other splicing factors and is involved in protein-nucleic acid and protein-protein interactions.Specifications
SLU7 | |
Polyclonal | |
Unconjugated | |
SLU7 | |
9G8; AU018913; D11Ertd730e; D3Bwg0878e; hSlu7; MGC9280; Pre-mRNA-splicing factor SLU7; Slu7; SLU7 homolog, splicing factor; SLU7 splicing factor homolog; SLU7 splicing factor homolog (S. cerevisiae); splicing factor; step II splicing factor SLU7; zinc knuckle motif containing | |
Rabbit | |
Protein A | |
RUO | |
10569 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O95391 | |
SLU7 | |
KLH conjugated synthetic peptide derived from human SLU7. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction